SY3H3D681038 Synthonix Corporation

(R)-Methyl 2-((2-chloro-5-nitropyrimidin-4-yl)(isopropyl)amino)butanoate

  • MDL number No MDL found
  • Cas Number : 946161-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56131.html

Safety Information

MSDS https://synthonix.com/56131.html
Availability details for
Quantity
  • Availability
  • Add to Cart