SY3H3D68103A Synthonix Corporation

4-(4-(Methylsulfonyl)phenyl)piperidine

  • Cas Number : 885274-65-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56166.html

Safety Information

MSDS https://synthonix.com/56166.html
Availability details for
Quantity
  • Availability
  • Add to Cart