SY3H3D68103B Synthonix Corporation

methyl4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 709648-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56183.html

Safety Information

MSDS https://synthonix.com/56183.html
Availability details for
Quantity
  • Availability
  • Add to Cart