SY3H3D68103C Synthonix Corporation

2-(4-Methylpiperidin-1-yl)aniline

  • MDL number MFCD04166381
  • Cas Number : 252758-94-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to light-yellow solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56190.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56190.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart