SY3H3D681040 Synthonix Corporation

(3-(4-Methylpiperazin-1-yl)phenyl)methanol

  • Cas Number : 123987-13-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56196.html

Safety Information

MSDS https://synthonix.com/56196.html
Availability details for
Quantity
  • Availability
  • Add to Cart