SY3H3D681048 Synthonix Corporation

Methyl adamantane-1-carboxylate

  • MDL number No MDL found
  • Cas Number : 711-01-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56219.html

Safety Information

MSDS https://synthonix.com/56219.html
Availability details for
Quantity
  • Availability
  • Add to Cart