SY3H3D68104D Synthonix Corporation

(S)-Methyl 2-((tert-butoxycarbonyl)amino)-3,3-dimethylbutanoate

  • MDL number No MDL found
  • Cas Number : 176504-88-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56321.html

Safety Information

MSDS https://synthonix.com/56321.html
Availability details for
Quantity
  • Availability
  • Add to Cart