SY3H3D681052 Synthonix Corporation

Methyl 4-(4-fluorophenyl)pyrimidine-2-carboxylate

  • Cas Number : 1401162-80-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/56482.html

Safety Information

MSDS https://synthonix.com/56482.html
Availability details for
Quantity
  • Availability
  • Add to Cart