SY3H3D681053 Synthonix Corporation

3-(4-Methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)benzoic acid

  • Cas Number : 641571-13-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56483.html

Safety Information

MSDS https://synthonix.com/56483.html
Availability details for
Quantity
  • Availability
  • Add to Cart