SY3H3D681055 Synthonix Corporation

2-Methylnaphtho[2,1-d]oxazole

  • MDL number No MDL found
  • Cas Number : 20686-65-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/56511.html

Safety Information

MSDS https://synthonix.com/56511.html
Availability details for
Quantity
  • Availability
  • Add to Cart