SY3H3D681058 Synthonix Corporation

Methyl 4-bromonaphthalene-1-carboxylate

  • MDL number No MDL found
  • Cas Number : 35615-97-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56533.html

Safety Information

MSDS https://synthonix.com/56533.html
Availability details for
Quantity
  • Availability
  • Add to Cart