SY3H3D681059 Synthonix Corporation

2-Methyl-10H-benzo[b]thieno[2,3-e][1,4]diazepin-4-amine hydrochloride

  • MDL number MFCD03085920
  • Cas Number : 138564-60-0
  • Molecular Weight : 265.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Yellow to Brownish Yellow solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/56566.html

Safety Information

signalword Warning
pcodes P261,P305,P351,P338
MSDS https://synthonix.com/56566.html
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart