SY3H3D68105D Synthonix Corporation

Methyl 2-(pyrrolidin-2-yl)benzoate hydrochloride

  • MDL number No MDL found
  • Cas Number : 1203681-57-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56597.html

Safety Information

MSDS https://synthonix.com/56597.html
Availability details for
Quantity
  • Availability
  • Add to Cart