SY3H3D681060 Synthonix Corporation

Methyl 8-amino-5,6,7,8-tetrahydronaphthalene-2-carboxylate hydrochloride

  • MDL number No MDL found
  • Cas Number : 1956309-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56602.html

Safety Information

MSDS https://synthonix.com/56602.html
Availability details for
Quantity
  • Availability
  • Add to Cart