SY3H3D681063 Synthonix Corporation

trans-4-(4-Methoxyphenyl)pyrrolidine-3-carboxylic acid hydrochloride

  • MDL number No MDL found
  • Cas Number : 1049978-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/56607.html

Safety Information

MSDS https://synthonix.com/56607.html
Availability details for
Quantity
  • Availability
  • Add to Cart