SY3H3D681065 Synthonix Corporation

(S)-Methyl 2-amino-3-(2,4-dimethylphenyl)propanoate hydrochloride

  • MDL number No MDL found
  • Cas Number : 1391571-15-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/56624.html

Safety Information

MSDS https://synthonix.com/56624.html
Availability details for
Quantity
  • Availability
  • Add to Cart