SY3H3D681066 Synthonix Corporation

Methyl 3-amino-3-(2,3-dimethoxyphenyl)propanoate hydrochloride

  • MDL number No MDL found
  • Cas Number : 1369494-59-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56625.html

Safety Information

MSDS https://synthonix.com/56625.html
Availability details for
Quantity
  • Availability
  • Add to Cart