SY3H3D681068 Synthonix Corporation

1-Methyl-5-(4-(trifluoromethoxy)phenyl)-1H-pyrrole-2-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 1864060-29-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/56669.html

Safety Information

MSDS https://synthonix.com/56669.html
Availability details for
Quantity
  • Availability
  • Add to Cart