SY3H3D68106A Synthonix Corporation

7-Methoxy-1-naphthylacetonitrile

  • MDL number MFCD08704309
  • Cas Number : 138113-08-3
  • Molecular Weight : 197.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: White - Slightly pale reddish yellow
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/56707.html

Safety Information

signalword Warning
pcodes P280,P305,P351,P338
MSDS https://synthonix.com/56707.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart