SY3H3D68106D Synthonix Corporation

2-(Methylsulfonyl)-10H-phenothiazine

  • MDL number No MDL found
  • Cas Number : 23503-68-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/56719.html

Safety Information

MSDS https://synthonix.com/56719.html
Availability details for
Quantity
  • Availability
  • Add to Cart