SY3H3D681074 Synthonix Corporation

2'-Methyl-[1,1'-biphenyl]-2-ol

  • MDL number No MDL found
  • Cas Number : 77897-02-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56777.html

Safety Information

MSDS https://synthonix.com/56777.html
Availability details for
Quantity
  • Availability
  • Add to Cart