SY3H3D68107F Synthonix Corporation

(2'-Methyl-[1,1'-biphenyl]-4-yl)boronic acid

  • Cas Number : 491595-36-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56839.html

Safety Information

MSDS https://synthonix.com/56839.html
Availability details for
Quantity
  • Availability
  • Add to Cart