SY3H3D681084 Synthonix Corporation

(3R,6S)-10-methoxy-6-methyl-8,11-dioxo-4-oxa-1,7-diazatricyclo[7.4.0.03,7]trideca-9,12-diene-12-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 1335210-24-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56855.html

Safety Information

MSDS https://synthonix.com/56855.html
Availability details for
Quantity
  • Availability
  • Add to Cart