SY3H3D681085 Synthonix Corporation

Methyl 2-(4-bromobenzoyl)-3-(methylamino)but-2-enoate

  • Cas Number : 1423699-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56874.html

Safety Information

MSDS https://synthonix.com/56874.html
Availability details for
Quantity
  • Availability
  • Add to Cart