SY3H3D681086 Synthonix Corporation

(5-(Methoxycarbonyl)-1-tosyl-1H-pyrrol-3-yl)boronic acid

  • MDL number No MDL found
  • Cas Number : 916177-00-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56877.html

Safety Information

MSDS https://synthonix.com/56877.html
Availability details for
Quantity
  • Availability
  • Add to Cart