SY3H3D681088 Synthonix Corporation

Methyl 3-(4-hydroxyphenyl)hex-4-ynoate

  • MDL number No MDL found
  • Cas Number : 865234-02-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56883.html

Safety Information

MSDS https://synthonix.com/56883.html
Availability details for
Quantity
  • Availability
  • Add to Cart