SY3H3D681089 Synthonix Corporation

2-(6-Methoxy-3,4-dihydronaphthalen-1-yl)acetic acid

  • MDL number No MDL found
  • Cas Number : 40154-29-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56884.html

Safety Information

MSDS https://synthonix.com/56884.html
Availability details for
Quantity
  • Availability
  • Add to Cart