SY3H3D68108C Synthonix Corporation

3-(1-Methyl-1,2,3,6-tetrahydropyridin-4-yl)-1H-pyrrolo[2,3-b]pyridine

  • MDL number No MDL found
  • Cas Number : 325975-67-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/56899.html

Safety Information

MSDS https://synthonix.com/56899.html
Availability details for
Quantity
  • Availability
  • Add to Cart