SY3H3D68108D Synthonix Corporation

6-METHOXY-1,2,3,4-TETRAHYDROCARBAZOLE

  • MDL number No MDL found
  • Cas Number : 13070-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56903.html

Safety Information

MSDS https://synthonix.com/56903.html
Availability details for
Quantity
  • Availability
  • Add to Cart