SY3H3D68108E Synthonix Corporation

Methyl 2-(5-methoxy-2-methyl-1H-indol-3-yl)acetate

  • MDL number No MDL found
  • Cas Number : 7588-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56920.html

Safety Information

MSDS https://synthonix.com/56920.html
Availability details for
Quantity
  • Availability
  • Add to Cart