SY3H3D68108F Synthonix Corporation

3-(2-Methylquinolin-1-ium-1-yl)propane-1-sulfonate

  • MDL number No MDL found
  • Cas Number : 51583-69-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56922.html

Safety Information

MSDS https://synthonix.com/56922.html
Availability details for
Quantity
  • Availability
  • Add to Cart