SY3H3D681091 Synthonix Corporation

Methyl (S)-4-Benzyl-5-oxomorpholine-3-carboxylate

  • MDL number No MDL found
  • Cas Number : 1235181-00-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56926.html

Safety Information

MSDS https://synthonix.com/56926.html
Availability details for
Quantity
  • Availability
  • Add to Cart