SY3H3D681092 Synthonix Corporation

8-Methoxy-5-methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole

  • Cas Number : 618910-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56943.html

Safety Information

MSDS https://synthonix.com/56943.html
Availability details for
Quantity
  • Availability
  • Add to Cart