SY3H3D681093 Synthonix Corporation

Methyl 4-cyclopentylbenzoate

  • MDL number No MDL found
  • Cas Number : 160598-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56973.html

Safety Information

MSDS https://synthonix.com/56973.html
Availability details for
Quantity
  • Availability
  • Add to Cart