SY3H3D681099 Synthonix Corporation

5-(4-Methylpiperazin-1-yl)-1H-indole

  • MDL number No MDL found
  • Cas Number : 412049-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57008.html

Safety Information

MSDS https://synthonix.com/57008.html
Availability details for
Quantity
  • Availability
  • Add to Cart