SY3H3D68109F Synthonix Corporation

Methyl 3-amino-1-benzylpyrrolidine-3-carboxylate

  • MDL number No MDL found
  • Cas Number : 145602-88-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57077.html

Safety Information

MSDS https://synthonix.com/57077.html
Availability details for
Quantity
  • Availability
  • Add to Cart