SY3H3D6810A2 Synthonix Corporation

2-(2-Methyl-2H-tetrazol-5-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

  • MDL number No MDL found
  • Cas Number : 1056039-83-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/57089.html

Safety Information

MSDS https://synthonix.com/57089.html
Availability details for
Quantity
  • Availability
  • Add to Cart