SY3H3D6810A8 Synthonix Corporation

2'-O-(2-Methoxyethyl)-guanosine

  • Cas Number : 473278-54-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57152.html

Safety Information

MSDS https://synthonix.com/57152.html
Availability details for
Quantity
  • Availability
  • Add to Cart