SY3H3D6810B2 Synthonix Corporation

3-(4-Methylpiperazin-1-ylmethyl)benzylamine

  • MDL number No MDL found
  • Cas Number : 515162-19-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57224.html

Safety Information

MSDS https://synthonix.com/57224.html
Availability details for
Quantity
  • Availability
  • Add to Cart