SY3H3D6810B5 Synthonix Corporation

Methyl 4-(3-(tert-butoxycarbonyl)ureido)piperidine-4-carboxylate

  • Cas Number : 1416351-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57251.html

Safety Information

MSDS https://synthonix.com/57251.html
Availability details for
Quantity
  • Availability
  • Add to Cart