SY3H3D6810B6 Synthonix Corporation

12-Methoxy-12-oxododecanoic acid

  • MDL number No MDL found
  • Cas Number : 3903-40-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57268.html

Safety Information

MSDS https://synthonix.com/57268.html
Availability details for
Quantity
  • Availability
  • Add to Cart