SY3H3D6810BE Synthonix Corporation

Methyl 4-(piperidin-4-yloxy)benzoate hydrochloride

  • MDL number No MDL found
  • Cas Number : 455323-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/57429.html

Safety Information

MSDS https://synthonix.com/57429.html
Availability details for
Quantity
  • Availability
  • Add to Cart