SY3H3D6810C0 Synthonix Corporation

N-Methyl-N-((3R,4R)-4-methylpiperidin-3-yl)-7H-pyrrolo[2,3-d]pyrimidin-4-amine hydrochloride

  • MDL number No MDL found
  • Cas Number : 1260590-51-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57444.html

Safety Information

MSDS https://synthonix.com/57444.html
Availability details for
Quantity
  • Availability
  • Add to Cart