SY3H3D6810C1 Synthonix Corporation

4-(3-Methoxybenzyl)piperidine hydrochloride

  • MDL number MFCD08669672
  • Cas Number : 149986-58-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57445.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57445.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart