SY3H3D6810C8 Synthonix Corporation

Methyl 5-(benzo[d][1,3]dioxol-5-yl)picolinate

  • MDL number No MDL found
  • Cas Number : 1799412-38-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/57518.html

Safety Information

MSDS https://synthonix.com/57518.html
Availability details for
Quantity
  • Availability
  • Add to Cart