SY3H3D6810C9 Synthonix Corporation

2-(4-Methylphenyl)benzothiazole

  • MDL number No MDL found
  • Cas Number : 16112-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/57521.html

Safety Information

MSDS https://synthonix.com/57521.html
Availability details for
Quantity
  • Availability
  • Add to Cart