SY3H3D6810CC Synthonix Corporation

2-Methyl-[1,1'-biphenyl]-3-carbaldehyde

  • Cas Number : 89951-60-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57567.html

Safety Information

MSDS https://synthonix.com/57567.html
Availability details for
Quantity
  • Availability
  • Add to Cart