SY3H3D6810D3 Synthonix Corporation

4'-(Methylsulfonyl)-[1,1'-biphenyl]-2-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 893736-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57583.html

Safety Information

MSDS https://synthonix.com/57583.html
Availability details for
Quantity
  • Availability
  • Add to Cart