SY3H3D6810D6 Synthonix Corporation

Methyl 4-methyl-6-phenylpicolinate

  • Cas Number : 119715-66-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57616.html

Safety Information

MSDS https://synthonix.com/57616.html
Availability details for
Quantity
  • Availability
  • Add to Cart