SY3H3D6810D8 Synthonix Corporation

Methyl 3-benzenesulfonamidobenzoate

  • MDL number No MDL found
  • Cas Number : 107922-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57624.html

Safety Information

MSDS https://synthonix.com/57624.html
Availability details for
Quantity
  • Availability
  • Add to Cart